accessibility menu, dialog, popup

UserName

Viewing profile as user:
SiFFFFFFLi

CAS RN 17347-95-4

IUPAC Name: lithium(1+) hexafluorosilanediuide
Synonyms: lithium(1+) hexafluorosilanediuide
Molecular Weight (g/mol): 149.02
Molecular Formula: F6LiSi
InChi Key: ZXUANORTLYQJEG-UHFFFAOYSA-N
SMILES: [Li+].F[Si--](F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results