accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 17348-76-4

IUPAC Name: 2-(2-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 2-(2-hydroxyphenyl)-2,3-dihydro-1-benzopyran-4-one
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: KZKWCKFDCPVDFJ-UHFFFAOYNA-N
SMILES: OC1=CC=CC=C1C1CC(=O)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results