CAS RN 17350-84-4
IUPAC Name:
2-amino-2-methyl-3-phenylpropanoic acid
Synonyms:
2-amino-2-methyl-3-phenylpropanoic acid
Molecular Weight (g/mol):
179.22
Molecular Formula:
C10H13NO2
InChi Key:
HYOWVAAEQCNGLE-UHFFFAOYNA-N
SMILES:
CC(N)(CC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
Sigma Aldrich (S)-Ru(Oac)2(Segphos™)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.