accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3C

CAS RN 17351-07-4

IUPAC Name: 1,2-diethyl cyclohexane-1,2-dicarboxylate
Synonyms: 1,2-diethyl cyclohexane-1,2-dicarboxylate
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H20O4
InChi Key: ZTUZDYWYNQDJKR-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCCC1C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results