accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNHNCH3OO

CAS RN 17354-14-2

IUPAC Name: 1,4-bis(butylamino)-9,10-dihydroanthracene-9,10-dione
Synonyms: oil blue 35 colouring agent CI 61554 (solvent blue 35)
Molecular Weight (g/mol): 350.46
Molecular Formula: C22H26N2O2
InChi Key: OCQDPIXQTSYZJL-UHFFFAOYSA-N
SMILES: CCCCNC1=CC=C(NCCCC)C2=C1C(=O)C1=CC=CC=C1C2=O
Color 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)