accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 173550-33-9

IUPAC Name: 3,5-bis(4-nitrophenoxy)benzoic acid
Synonyms: 3,5-bis(4-nitrophenoxy)benzoic acid
Molecular Weight (g/mol): 396.31
Molecular Formula: C19H12N2O8
InChi Key: JOSXKPZXMVHRKU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(OC2=CC=C(C=C2)[N+]([O-])=O)=CC(OC2=CC=C(C=C2)[N+]([O-])=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results