accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17356-09-1

IUPAC Name: 1-iodo-4-(propan-2-yl)benzene
Synonyms: 1-iodo-4-isopropylbenzene
Molecular Weight (g/mol): 246.09
Molecular Formula: C9H11I
InChi Key: PQJOSEVTIKYWLH-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(I)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results