CAS RN 173604-33-6
IUPAC Name:
4-(diphenylmethyl)-1,3-oxazolidin-2-one
Synonyms:
4-(diphenylmethyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol):
253.3
Molecular Formula:
C16H15NO2
InChi Key:
QEOCTJMBYZNEJH-UHFFFAOYNA-N
SMILES:
O=C1NC(CO1)C(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone, 97%
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone, 97%
CAS: 173604-33-6 Molecular Formula: C16H15NO2 Molecular Weight (g/mol): 253.301 MDL Number: MFCD01863565 InChI Key: QEOCTJMBYZNEJH-AWEZNQCLSA-N Synonym: r-4-benzhydryloxazolidin-2-one,r-+-4-diphenylmethyl-2-oxazolidinone,r-4-methyldiphenyl-2-oxazolidinone,4r-4-diphenylmethyl-1,3-oxazolidin-2-one,pubchem11686,4r-4-benzhydryloxazolidine-2-one,r-4-diphenylmethyl oxazolidin-2-one,2-oxazolidinone,4-diphenylmethyl-, 4r,2-oxazolidinone, 4-diphenylmethyl-, 4r,r-+-4-diphenylmethyl-2-oxazolidin PubChem CID: 9837924 IUPAC Name: (4R)-4-benzhydryl-1,3-oxazolidin-2-one SMILES: C1C(NC(=O)O1)C(C2=CC=CC=C2)C3=CC=CC=C3