accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOHN

CAS RN 17368-12-6

IUPAC Name: 2-chloro-1,4-dihydropyridin-4-one
Synonyms: 2-chloro-1H-pyridin-4-one
Molecular Weight (g/mol): 129.54
Molecular Formula: C5H4ClNO
InChi Key: VBEHFOMFHUQAOW-UHFFFAOYSA-N
SMILES: ClC1=CC(=O)C=CN1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)