accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1737-93-5

IUPAC Name: 3,5-dichloro-2,4,6-trifluoropyridine
Synonyms: 3,5-dichloro-2,4,6-trifluoropyridine
Molecular Weight (g/mol): 201.96
Molecular Formula: C5Cl2F3N
InChi Key: PKSORSNCSXBXOT-UHFFFAOYSA-N
SMILES: FC1=NC(F)=C(Cl)C(F)=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results