accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17377-95-6

IUPAC Name: N-benzyl-4-methoxyaniline
Synonyms: N-benzyl-4-methoxyaniline
Molecular Weight (g/mol): 213.28
Molecular Formula: C14H15NO
InChi Key: LIJJGMDKVVOEFT-UHFFFAOYSA-N
SMILES: COC1=CC=C(NCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results