accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOHOOH2NOO

CAS RN 1738-76-7

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl 2-aminoacetate
Synonyms: benzyl 2-aminoacetate; para-toluene sulfonate
Molecular Weight (g/mol): 337.39
Molecular Formula: C16H19NO5S
InChi Key: WJKJXKRHMUXQSL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.NCC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)