CAS RN 173963-93-4
IUPAC Name:
(2S)-3-(4-cyanophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms:
(2S)-3-(4-cyanophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol):
411.44
Molecular Formula:
C25H19N2O4
InChi Key:
JOPKKUTWCGYCDA-QHCPKHFHSA-M
SMILES:
[O-]C(=O)[C@H](CC1=CC=C(C=C1)C#N)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Sigma Aldrich Fmoc-Phe(4-Cf3)-Oh
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.