accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NNHOO

CAS RN 17400-34-9

IUPAC Name: hydrogen benzyl N-(3-aminopropyl)carbamate chloride
Synonyms: hydrogen benzyl N-(3-aminopropyl)carbamate chloride
Molecular Weight (g/mol): 244.72
Molecular Formula: C11H17ClN2O2
InChi Key: XKMBTMXQMDLSRB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)