accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1741-83-9

H3CSCH3

CAS RN 1741-83-9

IUPAC Name: 1-(methylsulfanyl)pentane
Synonyms: methyl pentyl sulfide
Molecular Weight (g/mol): 118.24
Molecular Formula: C6H14S
InChi Key: FOJGPFUFFHWGFQ-UHFFFAOYSA-N
SMILES: CCCCCSC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results