accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1742-95-6

IUPAC Name: 10-amino-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 4-amino-1,8-naphthalimide dfp 1 solvent yellow 85
Molecular Weight (g/mol): 212.21
Molecular Formula: C12H8N2O2
InChi Key: SSMIFVHARFVINF-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=C3C(=O)NC(=O)C(C=C1)=C23

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results