accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 174350-05-1

IUPAC Name: 1-ethoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene
Synonyms: 1-ethoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene
Molecular Weight (g/mol): 282.38
Molecular Formula: C17H24F2O
InChi Key: BOAHGIPRRKDVQY-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=C(F)C(F)=C(OCC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results