accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFTiHH

CAS RN 17439-11-1

IUPAC Name: titanium(4+) dihydrogen hexafluoride
Synonyms: hexafluorotitanic acid
Molecular Weight (g/mol): 163.87
Molecular Formula: F6H2Ti
InChi Key: PFSXARRIPPWGNC-UHFFFAOYSA-J
SMILES: [H+].[H+].[F-].[F-].[F-].[F-].[F-].[F-].[Ti+4]
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)