accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1744-07-6

IUPAC Name: 1,3-bis[4-(trifluoromethyl)phenyl]thiourea
Synonyms: 1,3-bis[4-(trifluoromethyl)phenyl]thiourea
Molecular Weight (g/mol): 364.31
Molecular Formula: C15H10F6N2S
InChi Key: ABYZUZQRUYXHCW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(NC(=S)NC2=CC=C(C=C2)C(F)(F)F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results