accessibility menu, dialog, popup

UserName

Viewing profile as user:
IIIISOOOO

CAS RN 1745-83-1

IUPAC Name: tetraiodo-3H-2,1λ⁶-benzoxathiole-1,1,3-trione
Synonyms: tetraiodo-2,1λ⁶-benzoxathiole-1,1,3-trione
Molecular Weight (g/mol): 687.75
Molecular Formula: C7I4O4S
InChi Key: UQDQTEHMZFWZSX-UHFFFAOYSA-N
SMILES: IC1=C(I)C(I)=C(I)C2=C1C(=O)OS2(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results