accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17450-56-5

IUPAC Name: ethyl (2E)-4-oxo-4-phenylbut-2-enoate
Synonyms: ethyl (2E)-4-oxo-4-phenylbut-2-enoate
Molecular Weight (g/mol): 204.23
Molecular Formula: C12H12O3
InChi Key: ACXLBHHUHSJENU-CMDGGOBGSA-N
SMILES: CCOC(=O)\C=C\C(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)