accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2N(S)PHOOHOClHOO

CAS RN 174575-17-8

IUPAC Name: (2S)-2-amino-3-[2'-chloro-5-(phosphonomethyl)-[1,1'-biphenyl]-3-yl]propanoic acid
Synonyms: (2S)-2-amino-3-[2'-chloro-5-(phosphonomethyl)-[1,1'-biphenyl]-3-yl]propanoic acid
Molecular Weight (g/mol): 369.74
Molecular Formula: C16H17ClNO5P
InChi Key: VBRJFXSFCYEZMQ-HNNXBMFYSA-N
SMILES: N[C@@H](CC1=CC(CP(O)(O)=O)=CC(=C1)C1=CC=CC=C1Cl)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results