accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPOOH2C

CAS RN 1746-03-8

IUPAC Name: ethenylphosphonate
Synonyms: vinylphosphonic acid
Molecular Weight (g/mol): 106.02
Molecular Formula: C2H3O3P
InChi Key: ZTWTYVWXUKTLCP-UHFFFAOYSA-L
SMILES: [O-]P([O-])(=O)C=C
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)