accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1746-25-4

IUPAC Name: 2,4,5-triiodo-1H-imidazole
Synonyms: 2,4,5-triiodoimidazole
Molecular Weight (g/mol): 445.77
Molecular Formula: C3HI3N2
InChi Key: HIDCNJIBRZBEPN-UHFFFAOYSA-N
SMILES: IC1=NC(I)=C(I)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results