CAS RN 1746-28-7
IUPAC Name:
1-bromo-4-(2-bromoethyl)benzene
Synonyms:
1-bromo-4-(2-bromoethyl)benzene
Molecular Weight (g/mol):
263.96
Molecular Formula:
C8H8Br2
InChi Key:
APTDRDYSJZQPPI-UHFFFAOYSA-N
SMILES:
BrCCC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Bromophenethyl bromide, 96%
4-Bromophenethyl bromide, 96%
CAS: 1746-28-7 Molecular Formula: C8H8Br2 Molecular Weight (g/mol): 263.96 InChI Key: APTDRDYSJZQPPI-UHFFFAOYSA-N Synonym: 1-bromo-4-2-bromoethyl benzene,4-bromophenethyl bromide,4-bromophenethylbromide,benzene,1-bromo-4-2-bromoethyl,4-bromo-1-2-bromoethyl benzene,pubchem16698,4-bromobenzylmethyl bromide,acmc-209e8m,1-bromo-4-2-bromoethyl ;benzene PubChem CID: 11010825 IUPAC Name: 1-bromo-4-(2-bromoethyl)benzene SMILES: C1=CC(=CC=C1CCBr)Br