accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONH2O

CAS RN 1746-77-6

IUPAC Name: propan-2-yl carbamate
Synonyms: isopropyl carbamate
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: OVPLZYJGTGDFNB-UHFFFAOYSA-N
SMILES: CC(C)OC(N)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)