CAS RN 17464-88-9
IUPAC Name:
1,3,4,6-tetrakis(methoxymethyl)-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
Synonyms:
1,3,4,6-tetrakis(methoxymethyl)-dihydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
Molecular Weight (g/mol):
318.33
Molecular Formula:
C12H22N4O6
InChi Key:
XGQJGMGAMHFMAO-UHFFFAOYSA-N
SMILES:
COCN1C2C(N(COC)C1=O)N(COC)C(=O)N2COC
1
–
1
of
1
results
1
1,3,4,6-Tetrakis(methoxymethyl)glycoluril 98.0+%, TCI America™
CAS: 17464-88-9 Molecular Formula: C12H22N4O6 Molecular Weight (g/mol): 318.33 MDL Number: MFCD00402438 InChI Key: XGQJGMGAMHFMAO-UHFFFAOYSA-N PubChem CID: 87125 IUPAC Name: 1,3,4,6-tetrakis(methoxymethyl)-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione SMILES: COCN1C2C(N(COC)C1=O)N(COC)C(=O)N2COC