accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17469-89-5

(S)OHONH3CCH3

CAS RN 17469-89-5

IUPAC Name: (2S)-2-(dimethylamino)-3-phenylpropanoic acid
Synonyms: (2S)-2-(dimethylamino)-3-phenylpropanoic acid
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: HOGIQTACRLIOHC-JTQLQIEISA-N
SMILES: CN(C)[C@@H](CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)