accessibility menu, dialog, popup

UserName

Viewing profile as user:
Si

CAS RN 1747-92-8

IUPAC Name: tribenzylsilyl
Synonyms: tribenzylsilyl
Molecular Weight (g/mol): 301.48
Molecular Formula: C21H21Si
InChi Key: QXZGXRIXJAVMTI-UHFFFAOYSA-N
SMILES: C([Si](CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)