accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17476-13-0

NHOHONOHOHTi

CAS RN 17476-13-0

IUPAC Name: 2-({2-[bis(2-hydroxyethyl)amino]ethyl}(2-hydroxyethyl)amino)ethan-1-ol titanium
Synonyms: tetrahydroxyethyl ethylenediamine titanium
Molecular Weight (g/mol): 284.18
Molecular Formula: C10H24N2O4Ti
InChi Key: AKPNQQCZXQKCHO-UHFFFAOYSA-N
SMILES: [Ti].OCCN(CCO)CCN(CCO)CCO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results