CAS RN 17477-29-1
CAS RN 17477-29-1
IUPAC Name:
chlorodimethylpropylsilane
Synonyms:
chlorodimethylpropylsilane
Molecular Weight (g/mol):
136.69
Molecular Formula:
C5H13ClSi
InChi Key:
HXVPUKPVLPTVCQ-UHFFFAOYSA-N
SMILES:
CCC[Si](C)(C)Cl
1
–
1
of
1
results
1
Chlorodimethylpropylsilane 99.0+%, TCI America™

CAS: 17477-29-1 Molecular Formula: C5H13ClSi Molecular Weight (g/mol): 136.694 MDL Number: MFCD00039301 InChI Key: HXVPUKPVLPTVCQ-UHFFFAOYSA-N Synonym: propyldimethylchlorosilane,chlorodimethylpropylsilane,n-propyldimethylchlorosilane,silane, chlorodimethylpropyl,dimethylpropylchlorosilane,chlorodimethyl propyl silane,chlorodimethyl-n-propylsilane PubChem CID: 87130 IUPAC Name: chloro-dimethyl-propylsilane SMILES: CCC[Si](C)(C)Cl