accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH2NNH2O

CAS RN 17481-27-5

IUPAC Name: 3-amino-4-methoxybenzamide
Synonyms: 3-amino-4-methoxybenzamide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: INCJNDAQNPWMPZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1N)C(N)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results