accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 174899-81-1

IUPAC Name: 1,3-dimethyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: 1,3-dimethylimidazolium; bistriflylimide anion
Molecular Weight (g/mol): 377.28
Molecular Formula: C7H9F6N3O4S2
InChi Key: SAYMDKMGIAANGQ-UHFFFAOYSA-N
SMILES: CN1C=C[N+](C)=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results