accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175135-14-5

IUPAC Name: 4-bromo-6-(trifluoromethyl)-1H-1,3-benzodiazole
Synonyms: 4-bromo-6-(trifluoromethyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 265.03
Molecular Formula: C8H4BrF3N2
InChi Key: HYTQERQCUFICAX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=C2N=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results