accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175135-19-0

IUPAC Name: 5-(morpholin-4-yl)-2-nitrobenzen-1-olate
Synonyms: 5-(morpholin-4-yl)-2-nitrobenzenolate
Molecular Weight (g/mol): 223.21
Molecular Formula: C10H11N2O4
InChi Key: XXIIDARBNMZFEH-UHFFFAOYSA-M
SMILES: [O-]C1=CC(=CC=C1[N+]([O-])=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results