accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175135-22-5

IUPAC Name: 2-(propylsulfanyl)pyridine-3-carboxylate
Synonyms: 2-(propylsulfanyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 196.24
Molecular Formula: C9H10NO2S
InChi Key: CSMDLVRDBYKTAH-UHFFFAOYSA-M
SMILES: CCCSC1=NC=CC=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results