accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175135-24-7

IUPAC Name: 2-(propylsulfanyl)pyridine-3-carbonyl chloride
Synonyms: 2-(propylsulfanyl)pyridine-3-carbonyl chloride
Molecular Weight (g/mol): 215.7
Molecular Formula: C9H10ClNOS
InChi Key: FTEUTJYYROCUBI-UHFFFAOYSA-N
SMILES: CCCSC1=C(C=CC=N1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results