accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNH2NFF

CAS RN 175135-32-7

IUPAC Name: 4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
Synonyms: 4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 212.22
Molecular Formula: C9H6F2N2S
InChi Key: NDCSJUJQMRFHEX-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC(F)=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results