accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175135-74-7

IUPAC Name: (3-chloro-4-fluorophenyl)hydrazine
Synonyms: (3-chloro-4-fluorophenyl)hydrazine
Molecular Weight (g/mol): 160.58
Molecular Formula: C6H6ClFN2
InChi Key: DUORBNTWCCJANU-UHFFFAOYSA-N
SMILES: NNC1=CC=C(F)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results