accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175136-79-5

IUPAC Name: 1-(3,5-dichlorophenyl)-1H-pyrrole-2-carbaldehyde
Synonyms: 1-(3,5-dichlorophenyl)pyrrole-2-carbaldehyde
Molecular Weight (g/mol): 240.08
Molecular Formula: C11H7Cl2NO
InChi Key: GNBDQGBCNPLAQK-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC(Cl)=C1)N1C=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results