accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175136-89-7

IUPAC Name: 1-(2-chloro-6-phenoxyphenyl)methanamine
Synonyms: 1-(2-chloro-6-phenoxyphenyl)methanamine
Molecular Weight (g/mol): 233.7
Molecular Formula: C13H12ClNO
InChi Key: SNTOZVXKDWQFEW-UHFFFAOYSA-N
SMILES: NCC1=C(OC2=CC=CC=C2)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results