accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFFFOOH

CAS RN 17515-74-1

IUPAC Name: 5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid
Synonyms: 5-methyl-2-(trifluoromethyl)furan-3-carboxylic acid
Molecular Weight (g/mol): 194.11
Molecular Formula: C7H5F3O3
InChi Key: NURNFMNHTHXTDS-UHFFFAOYSA-N
SMILES: CC1=CC(C(O)=O)=C(O1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results