accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSSCH3N

CAS RN 175202-48-9

IUPAC Name: methyl 4-cyano-5-(methylsulfanyl)thiophene-2-carboxylate
Synonyms: methyl 4-cyano-5-(methylsulfanyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 213.27
Molecular Formula: C8H7NO2S2
InChi Key: MJOQLNYXSSXZDJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(C#N)=C(SC)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results