accessibility menu, dialog, popup

UserName

Viewing profile as user:
SO

CAS RN 175203-58-4

IUPAC Name: 4-(2-phenylethynyl)thiophene-2-carbaldehyde
Synonyms: 4-(2-phenylethynyl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 212.27
Molecular Formula: C13H8OS
InChi Key: SAYKNBQCVYQQJF-UHFFFAOYSA-N
SMILES: O=CC1=CC(=CS1)C#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results