accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175204-79-2

IUPAC Name: carbamothioylmethyl 2,2-dimethylpropanoate
Synonyms: carbamothioylmethyl 2,2-dimethylpropanoate
Molecular Weight (g/mol): 175.25
Molecular Formula: C7H13NO2S
InChi Key: COULAOZTCJTHOX-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)OCC(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results