accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FNCS

CAS RN 175205-39-7

IUPAC Name: 4-fluoro-2-isothiocyanato-1-methylbenzene
Synonyms: 4-fluoro-2-isothiocyanato-1-methylbenzene
Molecular Weight (g/mol): 167.2
Molecular Formula: C8H6FNS
InChi Key: QVCYTKXGZLOVFA-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1N=C=S
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results