accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17524-05-9

IUPAC Name: dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 326.17
Molecular Formula: C10H14MoO6
InChi Key: SKMUJBBRXZPAJY-VGKOASNMSA-L
SMILES: O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (4)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)