accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175276-49-0

IUPAC Name: methyl 4-formyl-1,2,5-trimethyl-1H-pyrrole-3-carboxylate
Synonyms: methyl 4-formyl-1,2,5-trimethylpyrrole-3-carboxylate
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: NEDLMVCFWFSFNJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)N(C)C(C)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results