accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrIBr

CAS RN 175278-10-1

IUPAC Name: 1,3-dibromo-2-iodo-5-methylbenzene
Synonyms: 1,3-dibromo-2-iodo-5-methylbenzene
Molecular Weight (g/mol): 375.83
Molecular Formula: C7H5Br2I
InChi Key: VLVXYJATBIXIPF-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(I)C(Br)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results