accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175278-14-5

IUPAC Name: 4-bromo-2-(trifluoromethoxy)benzene-1-sulfonyl chloride
Synonyms: 4-bromo-2-(trifluoromethoxy)benzenesulfonyl chloride
Molecular Weight (g/mol): 339.51
Molecular Formula: C7H3BrClF3O3S
InChi Key: ZZCIRIWJHAZTIW-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(C=CC(Br)=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results